Skip to Content
Merck
All Photos(1)

Key Documents

96310

Sigma-Aldrich

Z-Hyp-OH

≥99.0% (T)

Synonym(s):

N-Cbz-Hydroxy-L-proline, Z-L-4-Hydroxyproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO5
CAS Number:
Molecular Weight:
265.26
Beilstein:
90295
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (T)

form

powder

optical activity

[α]20/D −54±1°, c = 2% in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

104-107 °C

application(s)

peptide synthesis

SMILES string

O[C@@H]1C[C@H](N(C1)C(=O)OCc2ccccc2)C(O)=O

InChI

1S/C13H15NO5/c15-10-6-11(12(16)17)14(7-10)13(18)19-8-9-4-2-1-3-5-9/h1-5,10-11,15H,6-8H2,(H,16,17)/t10-,11+/m1/s1

InChI key

WWVCWLBEARZMAH-MNOVXSKESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service