Skip to Content
Merck
All Photos(1)

Key Documents

930199

Sigma-Aldrich

Ni(COD)(TSO-Ph)

≥95%

Synonym(s):

(1,5-Cyclooctadiene)(2,3,4,5-tetraphenylthiophene 1-oxide)nickel(0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H32NiOS
Molecular Weight:
571.40
UNSPSC Code:
12352103
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

Application

Ni(COD)(TSO-Ph) is a bench-stable 18-electron nickel–olefin complex that can be used for a variety of reactions.


Reference

Structurally Diverse Bench-Stable Nickel(0) Pre-Catalysts: APractical Toolkit for In Situ Ligation Protocols

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Engle lab pioneers efficient catalytic methods, leveraging substrate directivity for streamlined organic synthesis and selective product formation.

Engle lab pioneers efficient catalytic methods, leveraging substrate directivity for streamlined organic synthesis and selective product formation.

Engle lab pioneers efficient catalytic methods, leveraging substrate directivity for streamlined organic synthesis and selective product formation.

Engle lab pioneers efficient catalytic methods, leveraging substrate directivity for streamlined organic synthesis and selective product formation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service