Skip to Content
Merck
All Photos(1)

Documents

926620

Sigma-Aldrich

Upconversion Nanoparticles

NaYF4:Yb,Er@NaYF4, Oil soluble core-shell, fluorescence λex 980, green light

Synonym(s):

UCNPs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

Quality Level

concentration

5 mg/mL

matrix active group

Oleic acid surface treatment

particle size

30 nm±5 nm

solubility

cyclohexane: dispersible

fluorescence

λex 980 (green)
λem 540 nm
λem 650 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Upconversion nanoparticles (ucnps), are the result of a unique optical process in which near-infrared (NIR) light excitation is converted into visible and/or UV emission. Upconversion (UC) utilizes sequential absorption of multiple photons utilizing both lifetime and real ladder-like energy levels of ions in a host lattice to produce a higher energy anti-Stokes luminescence.

This optical features of UCNPs, results in deep tissue penetration and minimal autofluorescence background, for a broad range of applications of UCNP in diagnostics and biomedical imaging systems.

Applications include:

  • Fluorescent microscopy
  • Deep-tissue bioimaging
  • Nanomedicine
  • Optogenetics
  • Security labelling
  • Volumetric display

Legal Information

Product of RuixiBiotechCo.Ltd

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Upconversion nanoparticles: design, nanochemistry, and applications in theranostics
Chen G., et al.
Chemical Reviews, 114, 5161-5214 (2014)
Engineering bright sub-10-nm upconverting nanocrystals for single-molecule imaging
Gargas, D. J. et al.
Nature Nanotechnology, 9, 300?305-300?305 (2014)
Advances in highly doped upconversion nanoparticles
Wen, S., Zhou, J., Zheng, K. et al.
Nature Communications, 9, 2415-2415 (2018)
Amplified stimulated emission in upconversion nanoparticles for super-resolution nanoscopy
Liu, Y. et al.
Nature, 543, 229?233-229?233 (2017)
Engineered Upconversion Nanoparticles for Resolving Protein Interactions inside Living Cells
Drees, C. et al.
Angewandte Chemie (International Edition in English), 55, 11668?11672-11668?11672 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service