Skip to Content
Merck
All Photos(1)

Key Documents

926396

Sigma-Aldrich

Solution-processable Blue TADF TB-P3Cz

≥95%

Synonym(s):

9′-(2,12-Di-tert-butyl-5,9-dioxa-13b-boranaphtho- [3,2,1-de]anthracen-7-yl)-9,9′′-diphenyl-9H,9′H,9′′H-3,3′:6′,3′′-tercarbazole, TADF Blue, TADF no host

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H56BN3O2
CAS Number:
Molecular Weight:
1030.07
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

PL: 460 nm (film)


ΔEST : 0.113 eV

blue light

Quality Level

Assay

≥95%

form

powder

color

yellow

solubility

THF: soluble
dichloromethane: soluble
toluene: soluble

Orbital energy

HOMO -4.88 eV 
LUMO -1.72 eV 

Application

Purely organic TADF emitters can recruit both singlet and triplet excitions for light emission and hence achieve 100% internal quantum efficiency (IQE). Additionally, purely organic TADF emitters avoid the problems associated with the use of heavy-metal-based organometallic complexes.
Bright, deep blue and stable emitters for OLEDs are grand challenge on OLED design at the moment. In this context, our TB-P3Cz is a promising TADF as solution-processable TADF blue emitter without the need of a host with a singlet–triplet energy gap (ΔEST) of 0.113 eV.
TADF blue materials can be investigated for their potential in organic light-emitting diodes (OLEDs). TADF emitters exhibit efficient utilization of both singlet and triplet excitons, resulting in enhanced device efficiency and reduced energy loss. It can is suitable in R&D studies of optoelectronic devices that require blue emission within the visible spectrum.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient blue thermally activated delayed fluorescence organic light emitting diodes based on tercarbazole donor and boron acceptor dyads.
D.Karthik et al.,
Journal of Material Chemistry C, 8, 2272-2279 (2020)
Color-Tunable Boron-Based Emitters Exhibiting Aggregation-Induced Emission and Thermally Activated Delayed Fluorescence for Efficient Solution-Processable Nondoped Deep-Blue to Sky-Blue OLEDs.
H.J. Kim et al.
Advanced Mat. (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service