Skip to Content
Merck
All Photos(1)

Key Documents

926167

Sigma-Aldrich

Chitosan glycidyl methacrylate

Degree of methacrylation ~20%

Synonym(s):

Chitosan acrylate, Chitosan methacrylate, Methacrylate grafted chitosan, Methyl methacrylate modified chitosan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C13H21NO7)n (C8H13NO5)m
UNSPSC Code:
12352201
NACRES:
NA.23

Quality Level

storage temp.

2-8°C

SMILES string

CO[C@H]1[C@@H](O)[C@@H](N)[C@H](O[C@H]2[C@@H](O)[C@@H](NC(C)=O)[C@H](C)O[C@@H]2CO)O[C@@H]1COCC(O)COC(C(C)=C)=O

Application

The unique physicochemical properties offer chitosan great potential in a range of biomedical applications such as tissue engineering, drug delivery vehicles, and enzyme immobilization for biosensing. Methacrylate functionalized chitosan is thermo/photo cross-linkable and used as a precursor for the fabrication of hydrogels in a wide range of biomedical applications including tissue engineering, 3D bioprinting, and drug and gene delivery. Chitosan-based material also has mucoadhesive properties and hence widely used in transmucosal therapeutics delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shefali Jaiswal et al.
Carbohydrate polymers, 211, 109-117 (2019-03-03)
A methyl methacrylate (MMA) modified chitosan (CS) conjugate (CSMMA) has been synthesized by a green method via Michael addition reaction between CS and MMA in ethanol. The synthesized conjugate was characterized by FT-IR, 1H NMR, X-ray diffraction spectrometry and SEM
Farnoosh Pahlevanzadeh et al.
Materials (Basel, Switzerland), 13(11) (2020-06-18)
Chitosan (CS) has gained particular attention in biomedical applications due to its biocompatibility, antibacterial feature, and biodegradability. Hence, many studies have focused on the manufacturing of CS films, scaffolds, particulate, and inks via different production methods. Nowadays, with the possibility
Panita Maturavongsadit et al.
ACS applied bio materials, 4(3), 2342-2353 (2022-01-12)
3D bioprinting has recently emerged as a very useful tool in tissue engineering and regenerative medicine. However, developing suitable bioinks to fabricate specific tissue constructs remains a challenging task. Herein, we report on a nanocellulose/chitosan-based bioink, which is compatible with
Chandra M Valmikinathan et al.
Soft matter, 8(6), 1964-1976 (2012-02-14)
Hydrogel based scaffolds for neural tissue engineering can provide appropriate physico-chemical and mechanical properties to support neurite extension and facilitate transplantation of cells by acting as 'cell delivery vehicles'. Specifically, in situ gelling systems such as photocrosslinkable hydrogels can potentially
Md Minhajul Islam et al.
Bioactive materials, 5(1), 164-183 (2020-02-23)
In recent years, there have been increasingly rapid advances of using bioactive materials in tissue engineering applications. Bioactive materials constitute many different structures based upon ceramic, metallic or polymeric materials, and can elicit specific tissue responses. However, most of them

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service