920681
Lithium 2-(3-(4-(tert-butoxycarbonyl)-1,4-diazepan-1-yl)pyrazin-2-yl)acetate
Synonym(s):
2-(3-(4-(tert-Butoxycarbonyl)-1,4-diazepan-1-yl)pyrazin-2-yl)acetate lithium salt, Rigid linker for PROTAC® development
About This Item
form
powder
Quality Level
reaction suitability
reagent type: linker
functional group
Boc
carboxylic acid
storage temp.
2-8°C
SMILES string
CC(C)(OC(N1CCCN(C2=NC=CN=C2CC([O-])=O)CC1)=O)C.[Li+]
InChI
1S/C16H24N4O4.Li/c1-16(2,3)24-15(23)20-8-4-7-19(9-10-20)14-12(11-13(21)22)17-5-6-18-14;/h5-6H,4,7-11H2,1-3H3,(H,21,22);/q;+1/p-1
InChI key
OICLFZXQYGPRFY-UHFFFAOYSA-M
Related Categories
Application
Similar linkers in this series:
920517
920525
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service