Skip to Content
Merck
All Photos(1)

Key Documents

911208

Sigma-Aldrich

1-Hydrosilatrane

≥95%

Synonym(s):

Hydrosilatrane, Silatrane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO3Si
CAS Number:
Molecular Weight:
175.26
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: reductant

mp

249-256 °C

storage temp.

−20°C

InChI

1S/C6H12NO3Si/c1-4-8-11-9-5-2-7(1)3-6-10-11/h1-6H2

InChI key

XBRVPWBNRAPVCC-UHFFFAOYSA-N

Application

1-Hydrosilatrane is an air- and moisture-stable hydride reducing reagent that exhibits high functional group compatibility. The Adler group has used 1-hydrosilatrane for the reduction of ketones and aldehydes as well as the direct reductive amination of ketones under mild conditions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silatrane as a Practical and Selective Reagent for the Reduction of Aryl Aldehydes to Benzylic Alcohols.
Skrypai V, et al.
European Journal of Organic Chemistry, 2016(12), 2207- 2211 (2016)
1-Hydrosilatrane: A Locomotive for Efficient Ketone Reductions.
Varjosaari E S, et al.
European Journal of Organic Chemistry, 2017(2), 229-232 (2017)
Vladislav Skrypai et al.
The Journal of organic chemistry, 84(9), 5021-5026 (2019-04-12)
The asymmetric direct reductive amination of prochiral ketones with aryl amines using 1-hydrosilatrane with a chiral Brønsted acid catalyst is reported. This is the first known example of chiral Brønsted acid-catalyzed asymmetric reductive amination using a silane as the hydride

Articles

As a versatile hydride reagent, 1-hydrosilatrane can be used in reduction reactions to synthesize alcohols, amines, their chiral counterparts, and esters from aldehydes/ketones.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service