Skip to Content
Merck
All Photos(2)

Key Documents

910813

Sigma-Aldrich

mPEG24-NHS

Synonym(s):

Methoxypolyethylene glycol acetic acid N-succinimidyl ester, mPEG NHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H107NO29
CAS Number:
Molecular Weight:
1258.44
MDL number:
UNSPSC Code:
43211915
NACRES:
NA.23

Assay

95% (HPLC)

form

solid

mol wt

average Mn 1,250 (by NMR)

reaction suitability

reagent type: chemical modification reagent
reactivity: amine reactive

color

white to off-white

Ω-end

NHS ester

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

Application

Polyethylene glycols (PEGs) are FDA approved hydrophilic polymers. They are non-toxic, non-immunogenic and non-antigenic, and are widely used in a variety of biomedical applications such as bioconjugation, drug delivery, surface functionalization, and tissue engineering. PEG-NHS is an amine reactive PEG that can react to amino groups of lysine(s) on proteins or other biologics; reaction occurs at room temperature in <1hr at pH 7-8. Stable linker between PEG and NHS ester.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service