Skip to Content
Merck
All Photos(1)

Key Documents

901092

Sigma-Aldrich

Aluminum-doped zinc oxide ink for spin coating/slot-die coating

Synonym(s):

AZO dispersion, AZO ink, Al-doped ZnO ink, Helios′Ink H-DZ41006 semiconductive ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
UNSPSC Code:
12352311
NACRES:
NA.23

description

Conductivity: 10-3-10-4 S/cm
Work function: 3.6 ± 0.1 eV

Quality Level

form

liquid (dispersion)

sheet resistance

10-20 Ω/sq (in combination with silver nanowires)

color

slightly yellow to clear

surface tension

23 mN/m±5 mN/m

viscosity

<4 mPa.s(20 °C)

capacity

≥0.9 % loading (Nanoparticles loading)

General description

Helios′Ink H-DZ41006 semiconductive ink developed for the printed electronics is particularly well suited for OPV and PV.
This ink shows great performance on spin coating and slot die coating and is compatible with various flexible substrates (Kapton®, PET, PET/ITO, etc.) but also glass/ITO.
The use of Helios′Ink H-DZ41006 semiconductive ink is greatly improving the devices performances by reducing the roughness of the TCE without a negative impact on the electrical conductivity and the transparency. Life time of the devices is also increased.
The use of Helios′Ink H-DZ41006 provides ETL layer with high performances. In addition, compared to standard ETL, the PCE not retained even with higher thickness.

Features and Benefits

Product benefits:
  • Easy deposition under atmospheric conditions (temperature and pressure).
  • Good optical performances (visible light transmission >90%).
  • Non CMR ink.
  • Compatible with ITO layer and Ag NWs layer.
  • Compatible with most flexible substrates.
  • Low drying temperature making printing onto flexible substrates possible.

Preparation Note

Typical processing guideline:
Can be homogenized for 5 minutes in an ultrasonic bath in order to get rid of any aggregates.
Drying conditions: Oven, IR oven, vacuum oven.
Clean-up solution: Ethanol/Acetone.

Legal Information

Product of Genes′Ink.
Kapton is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

58.1 °F

Flash Point(C)

14.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service