Skip to Content
Merck
All Photos(2)

Key Documents

900208

Sigma-Aldrich

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate)

greener alternative

dry re-dispersible pellets, high conductivity

Synonym(s):

Orgacon DRY5, PEDOT:PSS, Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

pellets

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

sheet resistance

<200 Ω/sq (by addition of 5% diethylene glycol)

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate) (PEDOT:PSS) (dry re-dispersible pellets) is a conductive polymer that is formed by electropolymerizing 3,4-ethylenedioxythiophene in a solution of poly(styrenesulfonate)(PSS). PEDOT is doped with positive ions and PSS with negative ions. It has the following properties that make it viable polymer in organic electronics:
  • low band gap
  • good optical properties
  • high conductivity
  • low redox potential
  • easy processing
  • tunable film forming ability

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product is used in energy conversion and storage, thus has been enhanced for energy efficiency. Click here for more information.

Application

Orgacon DRY5 pellets can be re-dispersed in organic solvents such as ethanol. The resulting dispersion can be used in coating or ink formulations targeted for different applications requiring high conductivity.
PEDOT:PSS can be used as an electrode material with high mobility for charge carriers. It can be used for a wide range of energy based applications such as organic photovoltaics (OPV), dye sensitized solar cells (DSSCs), organic light emitting diodes (OLEDs), supercapacitors and other biomedical based sensors.
PEDOT:PSS can be used as the active semiconductor layer in OFETs. It is commonly used as a hole transport layer in OPVs.
Virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend.

Legal Information

Orgacon is a trademark of Agfa-Gevaert N.V.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High efficiency, fully inkjet printed organic solar cells with freedom of design
Eggenhuisen TM, et al.
Applied Physics Letters, 3(14), 7255-7262 (2015)
Functionalized graphene/poly (3, 4-ethylenedioxythiophene): polystyrenesulfonate as counter electrode catalyst for dye-sensitized solar cells
Yue G, et al.
Energy, 54(8), 315-321 (2013)
Roll-to-Roll Slot-Die Coated Organic Photovoltaic (OPV) Modules with High Geometrical Fill Factors
Galagan Y, et al.
Energy Technology, 3(8), 834-842 (2015)
Stability of the interface between indium-tin-oxide and poly (3, 4-ethylenedioxythiophene)/poly (styrenesulfonate) in polymer light-emitting diodes.
De Jong MP, et al.
Applied Physics Letters, 77(14), 2255-2257 (2000)
Stability of polypyrrole and poly (3, 4-ethylenedioxythiophene) for biosensor application.
Yamato H, et al.
Journal of Electroanalytical Chemistry, 397(1-2), 163-170 (1995)

Articles

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service