Skip to Content
Merck
All Photos(1)

Key Documents

86849

Sigma-Aldrich

Tetrabutylammonium acetate

technical, ≥90% (T)

Synonym(s):

TBAAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OCOCH3)
CAS Number:
Molecular Weight:
301.51
Beilstein:
3599376
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (T)

form

powder

mp

95-98 °C (lit.)

SMILES string

CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C2H4O2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2(3)4/h5-16H2,1-4H3;1H3,(H,3,4)/q+1;/p-1

InChI key

MCZDHTKJGDCTAE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tetrabutylammonium acetate is an effective alternative for sodium acetate (NaOAc) due to its good solubility in organic solvents.

Application

Tetrabutylammonium acetate (TBAAc) is a good source of nucleophilic acetate ion for SN2 substitution reactions. It is commonly used to displace sulfonates and allylic halides to get corresponding acetates. Additionally, TBAAc can also be used as a mild, soluble base in Sonogashira reaction and Heck arylation.

Other Notes

Reagent for the epimerization of hydroxyl groups; base-molten salt for directing Heck-type reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Jeffery, M. David
Tetrahedron Letters, 39, 5751-5751 (1998)
Pd nanoparticle catalyzed Heck arylation of 1, 1-disubstituted alkenes in ionic liquids. Study on factors affecting the regioselectivity of the coupling process.
Calo V, et al.
Organometallics, 22(21), 4193-4197 (2003)
Tetrabutylammonium Acetate.
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Secondary deuterium kinetic isotope effects and the intervention of nonclassical ions in the solvolysis of exo-norborn-2-yl bromobenzene-p-sulfonate.
Maskill H,
Journal of the American Chemical Society, 98(26), 8482-8485 (1976)
Ligand-, copper-, and amine-free Sonogashira reaction of aryl iodides and bromides with terminal alkynes.
Urgaonkar S and Verkade JG
The Journal of Organic Chemistry, 69(17), 5752-5755 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service