Skip to Content
Merck
All Photos(1)

Key Documents

86730

Sigma-Aldrich

2,3,4,6-Tetra-O-benzyl-D-glucopyranose

≥98.0% (TLC)

Synonym(s):

2,3,4,6-Tetra-O-benzyl-glucopyranose, 2,3,4,6-Tetrakis-O-(phenylmethyl)-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H36O6
CAS Number:
Molecular Weight:
540.65
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

form

solid

optical activity

[α]20/D +49±2°, c = 2% in dioxane

mp

151-156 °C

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC1O[C@H](COCc2ccccc2)[C@@H](OCc3ccccc3)[C@H](OCc4ccccc4)[C@H]1OCc5ccccc5

InChI

1S/C34H36O6/c35-34-33(39-24-29-19-11-4-12-20-29)32(38-23-28-17-9-3-10-18-28)31(37-22-27-15-7-2-8-16-27)30(40-34)25-36-21-26-13-5-1-6-14-26/h1-20,30-35H,21-25H2/t30-,31-,32+,33-,34?/m1/s1

InChI key

OGOMAWHSXRDAKZ-BKJHVTENSA-N

Other Notes

Important D-glucopyranose derivative for glucosylations and other reactions ; Preparation of the α-glucopyranosyl chloride ; Synthesis of 1-C-α-D-glucopyranose derivatives

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Allevi et al.
Journal of the Chemical Society. Chemical Communications, 101-101 (1987)
A.A. Pavia et al.
Carbohydrate Research, 79, 79-79 (1980)
P. Allevi et al.
Journal of the Chemical Society. Chemical Communications, 1245-1245 (1987)
S. Koto et al.
Bulletin of the Chemical Society of Japan, 53, 1761-1761 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service