Skip to Content
Merck
All Photos(3)

Key Documents

86473

Sigma-Aldrich

α-Terpinene

≥95.0% (GC)

Synonym(s):

alpha-Terpinene, 1-Isopropyl-4-methyl-1,3-cyclohexadiene, p-Mentha-1,3-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1853379
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.478 (lit.)

bp

173-175 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)C1=CC=C(C)CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,6,8H,5,7H2,1-3H3

InChI key

YHQGMYUVUMAZJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Terpinene, a cyclic monoterpene commonly used as a fragrance compound, is found in various essential oils. It is responsible for the antioxidant activity of tea tree oil.

Application

α-Terpinene can be used as an organic reductant in the guaiacol deoxygenation to yield phenols using V2O5/Al2O3 catalyst.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanadium catalyzed guaiacol deoxygenation
Filley J and Roth C
J. Mol. Catal. A: Chem., 139(2-3), 245-252 (1999)
Conformational Space and Photochemistry of α-Terpinene
Marzec KM, et al.
The Journal of Physical Chemistry A, 114(17), 5526-5536 (2010)
D Djenane et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 17(6), 505-515 (2011-11-04)
Essential oils (EOs) extracted by hydrodistillation from leaf parts of Algerian Eucalyptus globulus, Myrtus communis and Satureja hortensis were analyzed by gas chromatography/mass spectrometry (GC/MS). The main components of EOs obtained were γ-terpinene (94.48%), 1,8-cineole (46.98%) and carvacrol (46.10%), respectively
Nabila Bouderdara et al.
Natural product communications, 6(1), 115-117 (2011-03-04)
The essential oil of aerial parts of Cachrys libanotis L. (Apiaceae) from east Algeria was extracted by hydrodistillation and analyzed by GC-FID and GC-MS. Thirty-one compounds were identified, the main components being germacrene-D (18.0%), gamma-terpinene (6.4%), p-cymene (5.5%), caryophyllene oxide
Wei-bin Yuan et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(7), 1067-1069 (2011-11-10)
To analysis the constituents of volatile oil from Fructus Auranti Immaturus by GC-MS. The volatile oil was extracted by steam distillation, then separated by capillary gas chromatography. The constituents of volatile oil were identified and their amount were determined by

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service