Skip to Content
Merck
All Photos(1)

Key Documents

860166

Sigma-Aldrich

2-Imidazolidone-4-carboxylic acid

95%

Synonym(s):

2-Oxo-4-imidazolidinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O3
CAS Number:
Molecular Weight:
130.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

184-186 °C (dec.) (lit.)

SMILES string

OC(=O)C1CNC(=O)N1

InChI

1S/C4H6N2O3/c7-3(8)2-1-5-4(9)6-2/h2H,1H2,(H,7,8)(H2,5,6,9)

InChI key

KZKRPYCBSZIQKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of human BACE-1 inhibitors

Additive for crystal nucleation and growth

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Devarakonda, et al.,
Journal of Crystal Growth, 204, 525-538 (1999)
Heuisul Park et al.
Bioorganic & medicinal chemistry letters, 18(9), 2900-2904 (2008-04-25)
We describe synthesis and evaluation of a series of cyclic urea derivatives with hydroxylethylamine isostere. Modification of P3, P1, and P2' and combination of SAR display a >100-fold increase in potency with good cellular activity (IC(50)=0.15microM) relative to the previously
S Samuels et al.
Pharmacology, biochemistry, and behavior, 24(1), 143-146 (1986-01-01)
DBA/2J mice were exposed in utero, between days 15-18 of gestation, to either of two enzyme inhibitors, previously shown to decrease blood-brain, large-neutral amino acid transport in adults: L-methionine-RS-sulfoximine and 2-imidazolidone-4-carboxylic acid. The young mice demonstrated persistently altered motor behavior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service