Skip to Content
Merck
All Photos(1)

Key Documents

82820

Sigma-Aldrich

3-Pyridinesulfonic acid

≥98.0% (T)

Synonym(s):

P3SA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO3S
CAS Number:
Molecular Weight:
159.16
Beilstein:
120822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

form

powder

impurities

≤2% water

mp

>300 °C (lit.)

functional group

sulfonic acid

SMILES string

OS(=O)(=O)c1cccnc1

InChI

1S/C5H5NO3S/c7-10(8,9)5-2-1-3-6-4-5/h1-4H,(H,7,8,9)

InChI key

DVECLMOWYVDJRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Pyridinesulfonic acid can be used in the synthesis of reaction intermediates, such as silver 3-pyridinesulfonate and 3-pyridinesulfonyl chloride.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly selective guest uptake in a silver sulfonate network imparted by a tetragonal to triclinic shift in the solid state.
Makinen S, et al.
Chemistry?A European Journal , 7(23), 5176-5182 (2001)
Symmetrical and unsymmetrical quadruply aza-bridged, closely interspaced, cofacial bis (5, 10, 15, 20-tetraphenylporphyrin) s. 2. Synthesis, characterization, and conformational effects of solvents.
Karaman R, et al.
Journal of the American Chemical Society, 114(12), 4889-4898 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service