Skip to Content
Merck
All Photos(2)

Key Documents

791555

Sigma-Aldrich

Titania paste, active opaque

Synonym(s):

Greatcell Solar® 18NR-AO Titania Paste, TiO2 paste

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

description

Crystal Structure: > 99% anatase (analysis carried out on starting material, prior to paste manufacture)

Quality Level

form

paste (cream)

concentration

27.0 wt. %

avg. part. size

20 nm (active)
≤450 nm (scatter)

viscosity

40000-55000 mPa.s(20 °C) (Analysis carried out with 20mm 4 degree cone/plate; 40 s-1))

SMILES string

[Ti](=O)=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

General description

Titania paste, active opaque is a precursor for titania that has anatase particles and larger sized anatase scatter particles, used to form a white coating on the fluorine doped tin oxide glass (FTO) substrate. It is used in a variety of electrochemical devices to improve electron mobility and power efficiency.

Application

Titania paste has a hierarchical pore structure which allows non-agglomerated loading on nanomaterials for the fabrication of dye sensitized solar cells with higher light absorption.
Use Active Opaque Titania Paste in applications that do not require transparency.
The dispersed scattering particles in Active Opaque Titania Paste formulation lead to enhanced DSC performance through the large surface/volume ratio of the sintered film.
Active Opaque Titania Paste has a blend of active anatase particles (~20nm) and larger anatase scatter particles (up to 450nm).
After drying; this paste must be fired at or above 500°C. This results in an opaque sintered layer with film thickness of 7-8μm for one printed layer and ~15μm for two layers; when using a 48T mesh.
This paste exhibits optimal rheological properties that provide good surface uniformity and contain organic binders specially formulated to provide versatile porosity suitable for a range of dye/electrolyte systems.
Storage: Store in the dark at 20°C

Legal Information

Product of Greatcell Solar® exclusive supplier of Sun2 products under license from EPFL.
Greatcell Solar® is a registered trademark of Greatcell Solar
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiazolo [5, 4-d] thiazole-based organic sensitizers with strong visible light absorption for transparent, efficient and stable dye-sensitized solar cells
Dessi A, et al.
Royal Society of Chemistry Advances, 5(41), 32657-32668 (2015)
Mesoporous titania hollow spheres applied as scattering layers in quantum dots sensitized solar cells
Xu X, et al.
Materials Chemistry and Physics, 136(2-3), 1060-1066 (2012)
Formation of size-tunable dandelion-like hierarchical rutile titania nanospheres for dye-sensitized solar cells
Lan C, et al.
Royal Society of Chemistry Advances, 3(2), 559-565 (2013)
Towards green, efficient and durable quasi-solid dye-sensitized solar cells integrated with a cellulose-based gel-polymer electrolyte optimized by a chemometric DoE approach
Bella F, et al.
Royal Society of Chemistry Advances, 3(36), 15993-16001 (2013)
Sol-gel based TiO2 paste applied in screen-printed dye-sensitized solar cells and modules
Hovcevar M, et al.
Journal of Industrial and Engineering Chemistry, 19(5), 1464-1469 (2013)

Articles

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service