Skip to Content
Merck
All Photos(1)

Documents

778877

Sigma-Aldrich

4-Azidobenzoic acid solution

~0.2 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

9-Azidobenzoic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
Beilstein:
1950876
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

form

liquid

concentration

~0.2 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

2-8°C

SMILES string

OC(C1=CC=C(N=[N+]=[N-])C=C1)=O

InChI

1S/C7H5N3O2/c8-10-9-6-3-1-5(2-4-6)7(11)12/h1-4H,(H,11,12)

InChI key

PQXPAFTXDVNANI-UHFFFAOYSA-N

General description

4-Azidobenzoic acid is an aromatic azide generally used in copper(I)-catalyzed azide-alkyne cycloaddition reactions.

Application

Review

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bao Li et al.
Regenerative biomaterials, 5(3), 159-166 (2018-06-27)
Photo-reactive poly(vinyl alcohol) (PRPVA) was synthesized by introduction of phenyl azido groups into poly(vinyl alcohol) (PVA) and applied for surface modification. PRPVA was grafted onto cell culture plate surface homogeneously or in a micropattern. Human mesenchymal stem cells (hMSCs) cultured
Aiping Zhu et al.
Biomaterials, 23(23), 4657-4665 (2002-09-27)
Chitosan (CS) was covalently immobilized onto polylactic acid (PLA) film surface using the photosensitive hetero-bifunctional crosslinking reagent, 4-azidobenzoic acid, which was previously bonded to chitosan by reaction between an acid group of the crosslinking reagent and a free amino group
Lu Sun et al.
Sheng wu yi xue gong cheng xue za zhi = Journal of biomedical engineering = Shengwu yixue gongchengxue zazhi, 23(2), 370-374 (2006-05-19)
Grafting of poly (ethylene glycol) (PEG) on the surface of polysulfone (PSF) sheets by simultaneous or sequential UV irradiation with 4-azidobenzoic acid as the photocoupler was carried out. Water contact angle measurements showed that there was a great improvement of
Stefan Müller et al.
Journal of materials chemistry. B, 8(3), 416-425 (2019-12-14)
Here, the surface of silicone elastomer was modified with photo-reactive gelatin bearing azidophenyl groups. Two types of gelatin were prepared: one by coupling with azidoaniline and the other by coupling with azidobenzoic acid. The silicone surface was hydrolyzed by oxygen
P Romero et al.
Journal of immunological methods, 171(1), 73-84 (1994-05-02)
A novel procedure is presented describing the induction of antigen-specific cytolytic T lymphocytes (CTL) in vivo, that uses as immunogen syngeneic Concanavalin A stimulated spleen cells expressing H-2Kd (Kd) molecules photocrosslinked with a photoreactive peptide derivative. The Kd restricted Plasmodium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service