Skip to Content
Merck
All Photos(1)

Key Documents

77411

Sigma-Aldrich

Peppermint oil

natural, from Mentha piperita L.

Synonym(s):

Mentha piperita

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

biological source

Mentha piperita L.

form

oil

quality

natural

refractive index

n20/D 1.46
n20/D 1.461 (lit.)

bp

215 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elise Herro et al.
Dermatitis : contact, atopic, occupational, drug, 21(6), 327-329 (2010-12-15)
PEPPERMINT (Mentha piperita) is a popular herb that can be used in numerous forms (ie, oil, leaf, leaf extract, and leaf water). Peppermint oil has the most uses, and use data on the oil are considered relevant to the leaf
Paul Enck et al.
European journal of gastroenterology & hepatology, 22(12), 1402-1411 (2011-03-11)
Numerous meta-analyses have recently assessed the overall clinical benefit of single therapy options and groups of therapies in the irritable bowel syndrome (IBS). By large, this should enable physicians to select from a number of therapy options available. We entered
Jacob F Smith et al.
Frontiers in physiology, 11, 80-80 (2020-03-07)
Osteoarthritis (OA) is a degenerative joint disease characterized by inflammatory degradation of articular cartilage and subchondral bone. Wogonin, a compound extracted from the plant Scutellaria baicalensis (colloquially known as skullcap), has previously been shown to have direct anti-inflammatory and antioxidative
M S Alam et al.
Mymensingh medical journal : MMJ, 22(1), 27-30 (2013-02-19)
Irritable bowel syndrome (IBS) is one of the most common functional gastrointestinal disorder which is associated with considerable sufferings of patient and Peppermint oil is volatile oil, its active principle is menthol-contain a cyclic monoterpine which has anti-spasmotic properties due
Naoki Hiki et al.
Digestive endoscopy : official journal of the Japan Gastroenterological Endoscopy Society, 24(2), 79-86 (2012-02-22)
Peppermint oil solution was found to be effective for reducing gastric spasm during upper gastrointestinal endoscopy. The aim of the present study was to assess whether the gastric peristalsis-suppressing effect is dose-dependently induced by L-menthol, the major constituent of peppermint

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service