Skip to Content
Merck
All Photos(1)

Documents

772313

Sigma-Aldrich

4-Methoxyphenyl β-D-glucopyranoside

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

133-172 °C

storage temp.

2-8°C

SMILES string

COc1ccc(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)cc1

InChI

1S/C13H18O7/c1-18-7-2-4-8(5-3-7)19-13-12(17)11(16)10(15)9(6-14)20-13/h2-5,9-17H,6H2,1H3/t9-,10-,11+,12-,13-/m1/s1

InChI key

SIXFVXJMCGPTRB-UJPOAAIJSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Aerts et al.
Biochimica et biophysica acta, 660(2), 317-324 (1981-08-13)
The interaction of alcohols in the hydrolysis of aryl beta-D-glucopyranosides and aryl beta-D-xylopyranosides by beta-D-glucosidase (beta-D-glucoside glucohydrolase, EC 3.2.1.21) from Stachybotrys atra has been investigated. The results constitute support for the presence of a glycosyl-enzyme intermediate, formed during the first
Z Zhang et al.
Carbohydrate research, 295, 41-55 (1996-12-13)
p-Methoxyphenyl (pMP) beta-D-glycopyranosides (Glc, Gal, GlcNPhth, GalNPhth, GlcNTroc, Gal beta 4Glc, Gal alpha 4Gal) were prepared from the corresponding 1-O-acetyl sugars in 79-90% yield, using boron trifluoride etherate as promoter. Treatment of the pMP glycosides with acyl chlorides or bromides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service