760919
Poly(ethylene glycol), 8 acetylene dendron, generation 2
average Mn 6,900
Synonym(s):
Polyethylene glycol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
description
Esterified with 8 acetylene groups
form
solid
mol wt
PEG average Mn 6,000
average Mn 6,900
feature
generation 2
no. Surface Groups
8
transition temp
Tm 47-52 °C
functional group
alkyne
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Dendrimers and dendrons are symmetrically branched polymer structures that possess a well-defined spatial distribution of functional groups. Dendrimers are chemically modified by attaching Poly ethylene glycol (PEG) molecule to increase its solubility and prolong circulation half-life. Clickable groups”, such as primary acetylenes, are introduced to the mono dispersed dendritic structures, using “click” chemistry.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biocompatible Dendritic Building Blocks for Advanced Biomedical Reasearch
Michael Malkoch and Andreas M Nystrom
Material Matters, 7(3), 47-49 null
Degradable Poly (ethylene glycol) hydrogels for 2 D and 3D cell culture
Andrea M Kasko
Material Matters, 6 (3), 67-75 null
Huiyuan Li et al.
Molecules (Basel, Switzerland), 18(8), 9797-9817 (2013-08-21)
Click chemistry is an efficient and chemoselective synthetic method for coupling molecular fragments under mild reaction conditions. Since the advent in 2001 of methods to improve stereochemical conservation, the click chemistry approach has been broadly used to construct diverse chemotypes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service