Skip to Content
Merck
All Photos(1)

Key Documents

75490

Sigma-Aldrich

DL-Ornithine monohydrochloride

≥99.0% (AT)

Synonym(s):

(±)-2,5-Diaminopentanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3CH(CO2H)(NH2) · HCl
CAS Number:
Molecular Weight:
168.62
Beilstein:
4153338
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (AT)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

~233 °C (dec.)

application(s)

peptide synthesis

SMILES string

Cl.NCCCC(N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H

InChI key

GGTYBZJRPHEQDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Ornithine monohydrochloride may be used as a starting material in the synthesis of (-)-(1-2H)putrescine dihydrochloride via enzymatic decarboxylation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Stereochemistry of the Enzymic Decarboxylation of l-Arginine and of l-Ornithine
Richards JC and Spenser ID
Canadian Journal of Chemistry, 60(22), 2810-2820 (1982)
Sherry Dadsetan et al.
Biochemical pharmacology, 85(1), 115-123 (2012-10-30)
Combined administration of ornithine and phenylacetate (OP) is proposed as a novel treatment of hyperammonemia and hepatic encephalopathy. Ornithine is believed to increase ammonia fixation into glutamine in muscle tissue and glutamine is subsequently thought to react with phenylacetate forming
Juan C Marini et al.
American journal of physiology. Endocrinology and metabolism, 303(11), E1348-E1353 (2012-10-18)
Citrulline is an amino acid synthesized in the gut and utilized for the synthesis of the conditionally essential amino acid arginine. Recently, the origin of the ornithine utilized for citrulline synthesis has become a matter of discussion. Multiple physiological factors
D Amelio et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 164(2), 356-362 (2012-11-06)
The Frank-Starling law is a fundamental property of the vertebrate myocardium which allows, when the end-diastolic volume increases, that the consequent stretch of the myocardial fibers generates a more forceful contraction. It has been shown that in the eel (Anguilla
Osama I Abd El-Salam et al.
Molecules (Basel, Switzerland), 17(12), 14510-14521 (2012-12-12)
A series of macrocyclic imides and Schiff-bases have been prepared via the cyclocondensation of pyridine-2,6-dicarbonyl dichloride (1) with L-ornithine methyl ester to give the corresponding macrocyclic bisester 2. Treatment of 2 with hydrazine hydrate gave macrocyclic bisacid hydrazide 3, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service