Skip to Content
Merck
All Photos(2)

Key Documents

752843

Sigma-Aldrich

Lithium carbonate

greener alternative

99.999% trace metals basis

Synonym(s):

Carbolithium, Carbonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2CO3
CAS Number:
Molecular Weight:
73.89
Beilstein:
3999191
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
powder

Quality Level

Assay

99.999% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤15.0 ppm Trace Metal Analysis

mp

618 °C (lit.)

density

2.11 g/cm3

greener alternative category

SMILES string

[Li+].[Li+].[O-]C([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

XGZVUEUWXADBQD-UHFFFAOYSA-L

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Lithium carbonate - 99.999% trace metal basis is a white powder with trace metal impurities ≤ 15.0 ppm. It is designed for application in industries and research fields that demand exceptional purity standards to achieve precise and reliable results in their processes and products.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium carbonate is one of the most important basic lithium salts, widely used in lithium-ion batteries, mainly used as a precursor for synthesizing lithium-ion battery cathode material. It is used to produce materials such as LCO(Lithium Cobalt Oxide), LMO(Lithium ion Manganese Oxide), LTO(Lithium Titanate Oxide), LFP, NMC111, NMC442, NMC532, NMC622 for Li-ion battery and those for other alkaline batteries. It can be used in research areas of therapies such as bipolar disorder, and certain neurological disorders. In the glass industry, lithium carbonate is employed in the production of specialized and optical glass. Additionally, it serves as a flux for making ductile ceramics, ceramic coatings used in metal preservation, and heat-resistant ceramic coatings. It is used as a deoxidizer in the process of industrial copper and nickel smelting; lithium can be used as a sulfur cleaner. It is also used in alloys with a variety of metals.

Features and Benefits

Exceptional Purity: The 99.999% purity minimizes contamination from trace metals, ensuring suitability for applications sensitive to even minute impurities. Consistent Performance: Ultra-high purity guarantees consistent performance across various applications, reducing variability and enhancing reliability. High Purity Standard: Ideal as a standard or reagent for trace metal analysis and high-precision analytical techniques, ensuring accurate and reliable results.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of lithium titanate (Li4Ti5O12) by addition of excess lithium carbonate (Li2CO3) in titanium dioxide (TiO2) xerogel
INTERNATIONAL JOURNAL OF TECHNOLOGY null
K E Morrison et al.
The Lancet. Neurology, 12(4), 339-345 (2013-03-05)
Lithium has neuroprotective effects in cell and animal models of amyotrophic lateral sclerosis (ALS), and a small pilot study in patients with ALS showed a significant effect of lithium on survival. We aimed to assess whether lithium improves survival in
N Alessi et al.
Journal of the American Academy of Child and Adolescent Psychiatry, 33(3), 291-304 (1994-03-01)
The use of lithium to treat child and adolescent psychiatric disorders is becoming more common. Since the publication of the report of The Committee on Biological Aspects of Child Psychiatry of the American Academy of Child Psychiatry in 1978, a
Andrea Contestabile et al.
The Journal of clinical investigation, 123(1), 348-361 (2012-12-04)
Down syndrome (DS) patients exhibit abnormalities of hippocampal-dependent explicit memory, a feature that is replicated in relevant mouse models of the disease. Adult hippocampal neurogenesis, which is impaired in DS and other neuropsychiatric diseases, plays a key role in hippocampal
Tom Bschor et al.
Journal of clinical psychopharmacology, 33(1), 38-44 (2013-01-02)
Studies of the 1970s and 1980s showed lithium monotherapy to be an effective treatment of acute unipolar major depressive disorder (MDD) and hence as a potential alternative to monoaminergic antidepressants.The objective was to conduct the first comparison of a lithium

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service