Skip to Content
Merck
All Photos(1)

Documents

750794

Sigma-Aldrich

Ethyl 2-aminooxazole-4-carboxylate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O3
CAS Number:
Molecular Weight:
156.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

135-140 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)c1coc(N)n1

InChI

1S/C6H8N2O3/c1-2-10-5(9)4-3-11-6(7)8-4/h3H,2H2,1H3,(H2,7,8)

InChI key

NBABLVASYFPOEV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service