Skip to Content
Merck
All Photos(1)

Documents

750034

Sigma-Aldrich

Hexylphosphonic acid

95%

Synonym(s):

n-Hexylphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15O3P
CAS Number:
Molecular Weight:
166.16
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

solid

mp

97-103 °C

SMILES string

CCCCCCP(O)(O)=O

InChI

1S/C6H15O3P/c1-2-3-4-5-6-10(7,8)9/h2-6H2,1H3,(H2,7,8,9)

InChI key

GJWAEWLHSDGBGG-UHFFFAOYSA-N

General description

Hexylphosphonic acid(HPA) is a phosphonic acid derivative that can be used as a capping agent and a surfactant.

Application

HPA may be used as a surfactant to disperse multi-walled carbon nanotubes(MWCNTs). It can be modify the surface of barium titanate(BaTiO3) which can be used in the development of 3D-printable dielectric paste.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viscoelastic properties of a 3D-Printable high-dielectric paste with surface-modified BaTiO3.
Park JY, et al.
Composites Science and Technology, 159(13), 225-231 (2018)
Aqueous dispersion of multiwall carbon nanotubes with phosphonic acid derivatives.
Oueiny C, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 493(13), 41-51 (2016)
Feng Li et al.
Journal of colloid and interface science, 549, 33-41 (2019-04-25)
The severe interface charge recombination caused by the large energy difference between perovskite material and carbon electrode significantly limits the further performance improvement of the all-inorganic perovskite solar cells (PSCs). We apply innovatively multilayer of quaternary Ag-In-Ga-S (AIGS) quantum dots
Towards new oligomesogenic phosphonic acids as stabilizers of nanoparticles colloids in nematic liquid crystals.
Prodanov MF, et al.
Synlett, 26(13), 1905-1910 (2015)
Adam Roberge et al.
The journal of physical chemistry letters, 8(17), 4055-4060 (2017-08-12)
Recently the addition of M

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service