Skip to Content
Merck
All Photos(1)

Key Documents

750018

Sigma-Aldrich

L-Arginine-13C6,15N4,2,3,3,4,4,5,5-d7 hydrochloride

99 atom % 13C, 98 atom % 15N, 98 atom % D, 95% (CP)

Synonym(s):

(S)-2-Amino-5-guanidinopentanoic acid-13C6,15N4,2,3,3,4,4,5,5-d7 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215N13C(=15NH)15NH(13CD2)313CD(15NH2)13COOH • HCl
Molecular Weight:
227.63
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
98 atom % 15N
98 atom % D

Assay

95% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

222 °C (dec) (lit.)

mass shift

M+17

SMILES string

[15NH2][13C]([15NH][13C]([2H])([2H])[13C]([2H])([2H])[13C]([2H])([2H])[13C@@](N)([2H])[13C](O)=O)=[15NH].Cl

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1/i1+1D2,2+1D2,3+1D2,4+1D,5+1,6+1,8+1,9+1,10+1;

InChI key

KWTQSFXGGICVPE-URLWBABPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Daniel Green et al.
Developmental cell, 46(5), 651-667 (2018-08-28)
Spermatogenesis requires intricate interactions between the germline and somatic cells. Within a given cross section of a seminiferous tubule, multiple germ and somatic cell types co-occur. This cellular heterogeneity has made it difficult to profile distinct cell types at different stages
Adrienne Niederriter Shami et al.
Developmental cell, 54(4), 529-547 (2020-06-07)
Spermatogenesis is a highly regulated process that produces sperm to transmit genetic information to the next generation. Although extensively studied in mice, our current understanding of primate spermatogenesis is limited to populations defined by state-specific markers from rodent data. As

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service