Skip to Content
Merck
All Photos(2)

Key Documents

737364

Sigma-Aldrich

4′-Methylbiphenyl-4-sulfonyl chloride

95%

Synonym(s):

4-(4-Methylphenyl)benzenesulfonyl chloride, 4-(p-Tolyl)benzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11ClO2S
CAS Number:
Molecular Weight:
266.74
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

solid

mp

105-110 °C

SMILES string

Cc1ccc(cc1)-c2ccc(cc2)S(Cl)(=O)=O

InChI

1S/C13H11ClO2S/c1-10-2-4-11(5-3-10)12-6-8-13(9-7-12)17(14,15)16/h2-9H,1H3

InChI key

DBYXACYHGMSPMQ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service