Skip to Content
Merck
All Photos(1)

Key Documents

729078

Sigma-Aldrich

3,4-Ethylenedithiothiophene

Synonym(s):

3,4-Ethylenedithiothiophene, 2,3-Dihydrothieno[3,4-b][1,4]dithiine, 3,4-Ethylenedisulfanylthiophene, EDTT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6S3
CAS Number:
Molecular Weight:
174.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

refractive index

n20/D 1.685

density

1.374 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C1CSc2cscc2S1

InChI

1S/C6H6S3/c1-2-9-6-4-7-3-5(6)8-1/h3-4H,1-2H2

InChI key

HPGNGICCHXRMIP-UHFFFAOYSA-N

General description

3,4-Ethylenedithiothiophene (EDTT) is a dithiin based conducting polymer that is a sulfur analog of 3,4-ethylendioxythiophene (EDOT) that is majorly used in optoelectronic and electrochemical devices.

Application

EDTT can be electrochemically oxidized in acetonitrile to form poly(EDTT) films on the surface of the electrodes, which can be used in the fabrication of organic light emitting diodes (OLEDs) and organic photovoltaics (OPVs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F

Flash Point(C)

105 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathieu, T.; et al.
Macromolecules, 38, 6806-6812 (2005)
Redox doping behaviour of poly (3, 4-ethylenedithiothiophene)-The counterion effect.
Domagala W, et al.
Optical Materials, 33(9), 1405-1409 (2011)
Functionalized 3, 4-ethylenedithiathiophenes (EDTTs) as building blocks for poly (3, 4-ethylenedithiathiophene)(PEDTT) derivatives.
Blanco R, et al.
Tetrahedron Letters, 49(13), 2056-2059 (2008)
Randriamahazaka, H.; Sini, G.; Tran Van, F.
The Journal of Physical Chemistry C, 111, 4553-4560 (2007)
Eduardo Damous Feijó et al.
Arquivos brasileiros de oftalmologia, 83(1), 1-4 (2019-10-31)
To determine the reliability of the endoscopic dye transit test for the prediction of functional success after dacryocystorhinostomy. A cross-sectional study was conducted with 50 patients who underwent external dacryocystorhinostomy Group or transcanalicular dacryocystorhinostomy Group and had anatomically patent ducts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service