Skip to Content
Merck
All Photos(1)

Documents

725242

Sigma-Aldrich

Bromoferrocene

Synonym(s):

1-Bromoferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9BrFe
CAS Number:
Molecular Weight:
264.93
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: iron
reagent type: catalyst

mp

26-32 °C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.Br[C]2[CH][CH][CH][CH]2

InChI

1S/C5H4Br.C5H5.Fe/c6-5-3-1-2-4-5;1-2-4-5-3-1;/h1-4H;1-5H;

InChI key

RFYQCXFGACWJED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service