Skip to Content
Merck
All Photos(1)

Documents

725005

Sigma-Aldrich

6-Aminobenzimidazole

95%

Synonym(s):

1H-1,3-Benzodiazol-6-amine, 3H-Benzoimidazol-5-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

163-168 °C

SMILES string

Nc1ccc2nc[nH]c2c1

InChI

1S/C7H7N3/c8-5-1-2-6-7(3-5)10-4-9-6/h1-4H,8H2,(H,9,10)

InChI key

WFRXSXUDWCVSPI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Trivulzio et al.
Pharmacological research communications, 20(11), 975-982 (1988-11-01)
Benzimidazole and some of its derivatives as 4-nitro and 5-nitro-benzimidazoles, 2-amino-, 4-amino- and 5-aminobenzimidazoles have been tested on gastric acid secretion in Shay-rats. Only 5-aminobenzimidazole decreased the gastric secretory process basal or stimulated by betazole. The antisecretory properties of 5-aminobenzimidazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service