Skip to Content
Merck
All Photos(3)

Documents

723622

Sigma-Aldrich

9H-Carbazole-9-(4-phenyl) boronic acid pinacol ester

95%

Synonym(s):

9-(4-(4,4,5,5-Tetramethy-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24BNO2
Molecular Weight:
369.26
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

solid

mp

165-173 °C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(cc2)-n3c4ccccc4c5ccccc35

InChI

1S/C24H24BNO2/c1-23(2)24(3,4)28-25(27-23)17-13-15-18(16-14-17)26-21-11-7-5-9-19(21)20-10-6-8-12-22(20)26/h5-16H,1-4H3

InChI key

AHDSYMVAUJZCOP-UHFFFAOYSA-N

General description

9H-Carbazole-9-(4-phenyl) boronic acid pinacol ester is a compound that is majorly used as an intermediate in electronic devices. Its molecular structure includes benzene rings, boronic acid pinacol ester and carbazole rings.

Application

9H-Carbazole-9-(4-phenyl) boronic acid pinacol can be used as a donor molecule in the formation of novel donor-π-acceptor dyes for electrochemical applications. It can also be used in the formation of organic light emitting diodes (OLEDs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theoretical and experimental study on spectra, electronic structure and photoelectric properties of three nature dyes used for solar cells
Ren P, et al.
Journal of Molecular Liquids, 247(3), 193-206 (2017)
Synthesis, photophysical and redox properties of the D-pi-A type pyrimidine dyes bearing the 9-phenyl-9H-carbazole moiety
Verbitskiy EV, et al.
Journal of Fluorescence, 25(3), 763-775 (2015)
Spectroscopic studies on 9H-carbazole-9-(4-phenyl) boronic acid pinacol ester by DFT method
Sas EB, et al.
Journal of Molecular Structure, 1118, 124-138 (2016)
New V-shaped 2, 4-di (hetero) arylpyrimidine push-pull systems: Synthesis, solvatochromism and sensitivity towards nitroaromatic compounds
Verbitskiy EV, et al.
Dyes and Pigments, 159(3), 35-44 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service