All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H11NO
CAS Number:
Molecular Weight:
89.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
96%
form
liquid
refractive index
n20/D 1.448
density
0.917 g/mL at 25 °C
functional group
amine
hydroxyl
storage temp.
2-8°C
SMILES string
CNCCCO
InChI
1S/C4H11NO/c1-5-3-2-4-6/h5-6H,2-4H2,1H3
InChI key
KRGXWTOLFOPIKV-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
180.0 °F - closed cup
Flash Point(C)
82.2 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service