Skip to Content
Merck
All Photos(1)

Key Documents

713570

Sigma-Aldrich

1,5-Cyclooctadiene{[dibenzyl((4R,5R)-5-methyl-2-phenyl-4,5-dihydro-4-oxazolyl)methyl]dicyclohexylphosphinite κN:κP}iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenyl)borate

97%

Synonym(s):

Ubaphox, [((4R,5R)-Cy2-Ubaphox)Ir(COD)]BARF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H70BF24IrNO2P
CAS Number:
Molecular Weight:
1731.35
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

optical purity

ee: ≥99%

SMILES string

[Ir+].C1CC=CCCC=C1.C[C@H]2OC(=N[C@H]2C(Cc3ccccc3)(Cc4ccccc4)OP(C5CCCCC5)C6CCCCC6)c7ccccc7.FC(F)(F)c8cc(cc(c8)C(F)(F)F)[B-](c9cc(cc(c9)C(F)(F)F)C(F)(F)F)(c%10cc(cc(c%10)C(F)(F)F)C(F)(F)F)c%11cc(cc(c%11)C(F)(F)F)C(F)(F)F

InChI

1S/C37H46NO2P.C32H12BF24.C8H12.Ir/c1-29-35(38-36(39-29)32-21-11-4-12-22-32)37(27-30-17-7-2-8-18-30,28-31-19-9-3-10-20-31)40-41(33-23-13-5-14-24-33)34-25-15-6-16-26-34;34-25(35,36)13-1-14(26(37,38)39)6-21(5-13)33(22-7-15(27(40,41)42)2-16(8-22)28(43,44)45,23-9-17(29(46,47)48)3-18(10-23)30(49,50)51)24-11-19(31(52,53)54)4-20(12-24)32(55,56)57;1-2-4-6-8-7-5-3-1;/h2-4,7-12,17-22,29,33-35H,5-6,13-16,23-28H2,1H3;1-12H;1-2,7-8H,3-6H2;/q;-1;;+1/b;;2-1-,8-7-;/t29-,35-;;;/m1.../s1

InChI key

BGPHLJWQZLLYML-XCHXSORBSA-N

Application

Catalyst for enantioselective hydrogenation of alkenes

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

sold in collaboration with Solvias AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Threonine-derived phosphinite-oxazoline ligands for the Ir-catalyzed enantioselective hydrogenation.
Menges, F. and Pfaltz, A.
Advanced Synthesis & Catalysis, 344, 40-44 (2002)
A New Class of Modular Phosphinite-Oxazoline Ligands: Ir-Catalyzed Enantioselective Hydrogenation of Alkenes We thank Dr. Martin Studer and Dr. Benoît Pugin (Solvias AG, Basel) for fruitful discussions, and Prof. Kevin Burgess (Texas A&M University) for preprints of related unpublished work. Financial support by the Swiss National Science Foundation is gratefully acknowledged.
Jörg Blankenstein et al.
Angewandte Chemie (International ed. in English), 40(23), 4445-4447 (2002-10-31)

Related Content

Andreas Pfaltz has a longstanding interest in the design of chiral ligands for asymmetric catalysis. The semicorrins developed in his group served as the prototype for an important family of nitrogen ligands, the bisoxazolines, which have found widespread use in catalytic asymmetric synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service