Skip to Content
Merck
All Photos(1)

Documents

705268

Sigma-Aldrich

Biotin-(ring-6,6-d2)

≥98 atom % D, ≥97% (CP)

Synonym(s):

D-Biotin-d2-(ring-6,6-d2), Bios II-(ring-6,6-d2), Coenzyme R-(ring-6,6-d2), Vitamin B7-(ring-6,6-d2), Vitamin B8-(ring-6,6-d2), Vitamin H-(ring-6,6-d2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D2H14N2O3S
CAS Number:
Molecular Weight:
246.32
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

≥98 atom % D

Quality Level

Assay

≥97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

white to off-white

mp

231-233 °C (lit.)

mass shift

M+2

shipped in

dry ice

storage temp.

−20°C
−20°C

SMILES string

[2H]C1([2H])S[C@@H](CCCCC(O)=O)[C@H]2NC(=O)N[C@@H]12

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1/i5D2

InChI key

YBJHBAHKTGYVGT-OXQHWPBHSA-N

General description

Biotin-(ring-6,6-d2), an isotope-labeled analog of biotin, is also known as vitamin H (vitamin B7). It is a water-soluble vitamin B essentially involved in the synthesis of fatty acids and in gluconeogenesis. Biotin is found in plants and animals and its deficiency potentially causes retardation in growth, alopecia and neurological impairment.

Application

Biotin-(ring-6,6-d2) is also known as coenzyme R-(ring-6,6-d2). This product is used as an internal analytical standard for the quantitation of biotin concentration in dietary supplements, fortified foods or within a complex biological matrix.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Factitious Graves disease due to biotin immunoassay interference-a case and review of the literature
Elston MS, et al.
The Journal of clinical endocrinology and metabolism, 101(9), 3251-3255 (2016)
Agar plate method using Lactobacillus plantarum for biotin determination in serum and urine
FUKUI T, et al.
J. Nutr. Sci. Vitaminol., 40(5), 491-498 (1994)
Magnetic resonance imaging assays for dimethyl sulfoxide effect on cancer vasculature
Cyran CC, et al.
Investigative Radiology, 43(5), 298-298 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service