Skip to Content
Merck
All Photos(1)

Key Documents

695122

Sigma-Aldrich

Acetamide

sublimed, 99%

Synonym(s):

Amide C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONH2
CAS Number:
Molecular Weight:
59.07
Beilstein:
1071207
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 65 °C)

Quality Level

Assay

99%

form

sublimed

bp

221 °C (lit.)

mp

78-80 °C (lit.)

functional group

amide

SMILES string

CC(N)=O

InChI

1S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)

InChI key

DLFVBJFMPXGRIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristian Varela et al.
Chemistry & biology, 19(4), 498-506 (2012-04-24)
Mycolic acids are vital components of the cell wall of the tubercle bacillus Mycobacterium tuberculosis and are required for viability and virulence. While mycolic acid biosynthesis is studied extensively, components involved in mycolate transport remain unidentified. We investigated the role
A E Alves et al.
Reproduction in domestic animals = Zuchthygiene, 47(6), 1003-1008 (2012-03-06)
This study was undertaken to compare cryotolerance, in terms of viability and resumption of meiosis after warming and culture (24 and 48 h), of ex situ (isolated) and in situ (enclosed in the ovarian tissue) feline cumulus-oocyte complexes (COCs) vitrified
L Wang et al.
The journal of physical chemistry. B, 115(13), 3713-3724 (2011-03-17)
Infrared (IR) spectroscopy of the amide I band has been widely utilized for the analysis of peptides and proteins. Theoretical modeling of IR spectra of proteins requires an accurate and efficient description of the amide I frequencies. In this paper
Asal Fallah-Tafti et al.
European journal of medicinal chemistry, 46(10), 4853-4858 (2011-08-20)
KX2-391 (KX-01/Kinex Pharmaceuticals), N-benzyl-2-(5-(4-(2-morpholinoethoxy)phenyl)pyridin-2-yl)acetamide, is a highly selective Src substrate binding site inhibitor. To understand better the role of pyridine ring and N-benzylsubstitution in KX2-391 and establish the structure-activity relationship, a number of N-benzyl substituted (((2-morpholinoethoxy)phenyl)thiazol-4-yl)acetamide derivatives containing thiazole instead
E C B Silva et al.
Animal reproduction science, 132(3-4), 155-158 (2012-06-26)
The aim was to assess the in vitro effect of glycerol, ethylene glycol or acetamide on frozen-thawed ram spermatozoa. Aliquots of each sixteen ejaculates from four rams of the Morada Nova breed were diluted in Tris-egg yolk with glycerol (5%)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service