Skip to Content
Merck
All Photos(3)

Key Documents

683183

Sigma-Aldrich

CARAC

Umicore, 98%

Synonym(s):

Dicarbonyl-acetylacetonato-rhodium(I), (Acetylacetonato)dicarbonylrhodium(I), Dicarbonylrhodium(I) 2,4-pentanedionate, Rh(CO)2acac, Rhodium(I) dicarbonyl acetylacetonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rh(CO)2(C5H7O2)
CAS Number:
Molecular Weight:
258.03
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

mp

154-156 °C (lit.)

SMILES string

[C-]#[O+].[C-]#[O+].CC(=O)\C=C(\C)O[Rh]

InChI

1S/C5H8O2.2CO.Rh/c1-4(6)3-5(2)7;2*1-2;/h3,6H,1-2H3;;;/q;;;+1/p-1/b4-3-;;;

InChI key

BZCAWKOPWNIDOC-FGSKAQBVSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Umicore Precatalysts for Asymmetric and Cross-Coupling Catalysis

Catalyst for various carbonylation reactions, silylcarbocyclizations,conjugate additions to enones, carbamoylstannation,and reduction of aromatic nitro compounds

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Bonafoux et al.
Organic letters, 3(9), 1303-1305 (2001-05-12)
[reaction in text] Desymmetrization of dimethylsilyloxyalkadiynes (1) by Rh-catalyzed intramolecular silylformylation affords 5-exo-(formylmethylene)oxasilacyclopentanes 2 in high yields. Novel sequential double silylformylation of 1a also provides desymmetrization, giving 3-(3-silyl-2-formylprop-2-enyl)-5-exo-(formylmethylene)oxasilacyclopentanes 4 in excellent yields. Reduction of 2a and 4 with NaBH(4) gives
Ojima, I.; Lee, S.-Y.
Journal of the American Chemical Society, 122, 2385-2385 (2000)
Breit, B.; Seiche, W.,
Synthesis, 1-1 (2001)
Andrei V Pakoulev et al.
The journal of physical chemistry. A, 112(28), 6320-6329 (2008-06-25)
Triply vibrationally enhanced four-wave mixing spectroscopy is employed to observe vibrational coherence transfer between the asymmetric and symmetric CO-stretching modes of rhodium(I) dicarbonyl acetylacetonate (RDC). Coherence transfer is a nonradiative transition of a coherent superposition of quantum states to a
K A Merchant et al.
Physical review letters, 86(17), 3899-3902 (2001-05-01)
2D spectrally resolved ultrafast (<200 fs) IR vibrational echo experiments were performed on Rh(CO)(2)acac [(acetylacetonato)dicarbonylrhodium (I)]. The 2D spectra display features that reflect the 0-1 and 1-2 transitions and the combination band transition of the symmetric (S) and antisymmetric (A)

Articles

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service