Skip to Content
Merck
All Photos(2)

Key Documents

67550

Sigma-Aldrich

1,3,5-O-Methylidyne-myo-inositol

≥99.0% (HPLC)

Synonym(s):

myo-Inositol monoorthoformate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O6
CAS Number:
Molecular Weight:
190.15
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (HPLC)

form

solid

mp

260 °C (dec.) (lit.)
≥270 °C (dec.)

SMILES string

O[C@@H]1C2O[C@H]3O[C@@H]1[C@H](O)C(O3)[C@@H]2O

InChI

1S/C7H10O6/c8-1-4-2(9)6-3(10)5(1)12-7(11-4)13-6/h1-10H/t1-,2-,3-,4+,5?,6?,7-/m0/s1

InChI key

WJJFNSGWGPSKAO-XEKWHFIDSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Building block for the synthesis of inositol phosphates

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.K. Chung et al.
Chemical Communications (Cambridge, England), 163-163 (1996)
A.M. Riley et al.
The Journal of Organic Chemistry, 60, 4970-4970 (1995)
P. Andersch et al.
Tetrahedron Asymmetry, 7, 349-349 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service