Skip to Content
Merck
All Photos(1)

Documents

674737

Sigma-Aldrich

(R)-3,3′-Bis(triphenylsilyl)-1,1′-bi-2-naphthol

96%

Synonym(s):

(R)-3,3′-Bis(triphenylsilyl)-1,1′-binaphthalene-2,2′-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H42O2Si2
CAS Number:
Molecular Weight:
803.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

optical activity

[α]20/D +114°, c = 1 in chloroform

mp

103-145 °C

SMILES string

Oc1c(cc2ccccc2c1-c3c(O)c(cc4ccccc34)[Si](c5ccccc5)(c6ccccc6)c7ccccc7)[Si](c8ccccc8)(c9ccccc9)c%10ccccc%10

InChI

1S/C56H42O2Si2/c57-55-51(59(43-25-7-1-8-26-43,44-27-9-2-10-28-44)45-29-11-3-12-30-45)39-41-23-19-21-37-49(41)53(55)54-50-38-22-20-24-42(50)40-52(56(54)58)60(46-31-13-4-14-32-46,47-33-15-5-16-34-47)48-35-17-6-18-36-48/h1-40,57-58H

InChI key

STBZSRVMGWTCOU-UHFFFAOYSA-N

Application

(R)-3,3′-Bis(triphenylsilyl)-1,1′-bi-2-naphthol reacts with trimethylaluminum to form a chiral organoaluminum reagent, which can catalyze the asymmetric Diels-Alder reaction of cyclopentadiene and methyl acrylate to form the corresponding Diels-Alder adduct.
Precursor to a chiral Bronsted acid (674745) used to catalyze an enantioselective aza Diels-Alder reaction providing bicyclic lactams. Rare earth metal complexes of this chiral binapthol catalyze an intramolecular hydroamination of amino olefins leading to chiral pyrrolidines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Diels-Alder Reaction of Cyclopentadiene and Methyl Acrylate Catalyzed by Chiral Organoaluminum Reagents.
Bulletin of the Chemical Society of Japan, 65(12), 3501-3503 (1992)
Denis V Gribkov et al.
Journal of the American Chemical Society, 128(11), 3748-3759 (2006-03-16)
Chiral 3,3'-bis(trisarylsilyl)-substituted binaphtholate rare earth metal complexes (R)-[Ln{Binol-SiAr3}(o-C6H4CH2NMe2)(Me2NCH2Ph)] (Ln = Sc, Lu, Y; Binol-SiAr3 = 3,3'-bis(trisarylsilyl)-2,2'-dihydroxy-1,1'-binaphthyl; Ar = Ph (2-Ln), 3,5-xylyl (3-Ln)) and (R)-[La{Binol-Si(3,5-xylyl)3}{E(SiMe3)2}(THF)2] (E = CH (4a), N (4b)) are accessible via facile arene, alkane, and amine elimination. They
Hua Liu et al.
Organic letters, 8(26), 6023-6026 (2006-12-15)
[Structure: see text] The first chiral Brønsted acid-catalyzed asymmetric direct aza hetero-Diels-Alder reaction has been described. The phosphoric acids, prepared from binol and H8-binol derivatives, have shown catalytic ability for the reaction of cyclohexenone with N-PMP-benzaldimine. A chiral phosphoric acid

Articles

We present an article concerning BINOL and Derivatives.

Related Content

The Yamamoto group has employed several applications in catalysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service