Skip to Content
Merck
All Photos(1)

Key Documents

671444

Sigma-Aldrich

1,3-Dimethylimidazolium dimethyl phosphate

≥98.0% (HPLC)

Synonym(s):

[MMIM][DMP]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15N2O4P
CAS Number:
Molecular Weight:
222.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

impurities

≤1.0% water

functional group

phosphate

SMILES string

COP([O-])(=O)OC.Cn1cc[n+](C)c1

InChI

1S/C5H9N2.C2H7O4P/c1-6-3-4-7(2)5-6;1-5-7(3,4)6-2/h3-5H,1-2H3;1-2H3,(H,3,4)/q+1;/p-1

InChI key

GSGLHYXFTXGIAQ-UHFFFAOYSA-M

General description

1,3-Dimethylimidazolium dimethyl phosphate is an imidazolium-based phosphoric ionic liquid that can be prepared by reacting 1-methylimidazole with trimethyl phosphate. The pretreatment of sugarcane bagasse with [MMIM][DMP] yields glucose and xylose via enzymatic saccharification.

Application

1,3-Dimethylimidazolium dimethyl phosphate ([MMIM][DMP]) can be used as a catalyst and a solvent for the preparation of ethyl 2-cyano-3-phenylpropenoate from benzaldehyde and ethyl cyanoacetate via Knoevenagel condensation. It can also be used to produce bioethanol from sugarcane bagasse by enzymatic hydrolysis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Extractive desulfurization of gasoline using imidazolium-based phosphoric ionic liquids.
Nie Y, et al.
Energy and Fuels, 20(5), 2083-2087 (2006)
Imidazolium dialkylphosphates-a class of versatile, halogen-free and hydrolytically stable ionic liquids.
Kuhlmann E, et al.
Green Chemistry, 9(3), 233-242 (2007)
Dialkylimidazolium dimethyl phosphates as solvents and catalysts for the Knoevenagel condensation reaction.
Zicmanis A and Anteina L.
Tetrahedron Letters, 55(12), 2027-2028 (2014)
Samaneh Bahrani et al.
Bioresource technology, 185, 411-415 (2015-03-26)
In this study, an imidazolium-based ionic liquid (IL), 1,3-dimethylimidazolium dimethyl phosphate ([Mmim][DMP]), was applied for pretreating sugarcane bagasse to produce bioethanol. The main goal of this study was to investigate the feasibility of bagasse pretreatment with this IL, and to
Major improvement in the rate and yield of enzymatic saccharification of sugarcane bagasse via pretreatment with the ionic liquid 1-ethyl-3-methylimidazolium acetate ([Emim][Ac]).
da Silva ASA, et al.
Bioresource Technology, 102(22), 10505-10509 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service