Skip to Content
Merck
All Photos(1)

Key Documents

668478

Sigma-Aldrich

(+)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]ethane

kanata purity

Synonym(s):

(R,R)-Et-BPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36P2
CAS Number:
Molecular Weight:
314.43
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.5249

bp

104-106 °C/0.05 mmHg

density

0.939 g/mL at 25 °C

SMILES string

CC[C@@H]1CC[C@@H](CC)P1CCP2[C@H](CC)CC[C@H]2CC

InChI

1S/C18H36P2/c1-5-15-9-10-16(6-2)19(15)13-14-20-17(7-3)11-12-18(20)8-4/h15-18H,5-14H2,1-4H3/t15-,16-,17-,18-/m1/s1

InChI key

QOLRLVPABLMMKI-BRSBDYLESA-N

Application

(+)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]ethane can be used as a reactant to prepare:
  • Chromium diphosphine chloride complex which is employed as a catalyst for chemoselective oligomerization reaction.
  • α-Arylpyrrolidines by Suzuki-Miyaura cross-coupling and enantioselective copper-catalyzed intramolecular hydroamination reactions.

It can also be used as a catalyst in the enantioselective preparation of (perfluoroalkyl)butenyldiketones via cross Rauhut-Currier reaction of β-perfluoroalkylenones and vinyl ketones.

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphine-Catalyzed Asymmetric Intermolecular Cross Rauhut-Currier Reaction of β-Perfluoroalkyl-Substituted Enones and Vinyl Ketones
T Mengna, et al.
advanced synthesis and catalysis, 359(19), 3347-3353 (2017)
Phospholane-Based Ligands for Chromium-Catalyzed Ethylene Tri-and Tetramerization
Boelter SD, et al.
Organometallics, 39(7), 976-987 (2020)
Catalytic Asymmetric Synthesis of α-Arylpyrrolidines and Benzo-fused Nitrogen Heterocycles
Dai X-J, et al.
Angewandte Chemie (International Edition in English), 131(11), 3445-3449 (2019)
Phosphine-Catalyzed Asymmetric Intermolecular Cross Rauhut-Currier Reaction of β-Perfluoroalkyl-Substituted Enones and Vinyl Ketones
T Mengna, et al.
Advanced Synthesis & Catalysis, 359(19), 3347-3353 (2017)

Articles

Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.

Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.

Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.

Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service