668192
(S)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4- a′]dinaphthalen-4-yl)dimethylamine
97%
Synonym(s):
(S)-MonoPhos®
About This Item
Recommended Products
Assay
97%
form
solid
mp
190 °C (dec.)
functional group
amine
SMILES string
[p]1([o]c2c(c4c([o]1)ccc5c4cccc5)c3c(cc2)cccc3)N(C)C
InChI
1S/C22H18NO2P/c1-23(2)26-24-19-13-11-15-7-3-5-9-17(15)21(19)22-18-10-6-4-8-16(18)12-14-20(22)25-26/h3-14H,1-2H3
InChI key
QCHAVHXSBZARBO-UHFFFAOYSA-N
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.
DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.
DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.
DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service