658820
4-Bromo-2,6-dichloroaniline
97%
Synonym(s):
(4-Bromo-2,6-dichlorophenyl)amine, 2,6-Dichloro-4-bromoaniline, 4-Bromo-2,6-dichloro-benzenamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H4BrCl2N
CAS Number:
Molecular Weight:
240.91
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
83-87 °C
functional group
bromo
chloro
SMILES string
Nc1c(Cl)cc(Br)cc1Cl
InChI
1S/C6H4BrCl2N/c7-3-1-4(8)6(10)5(9)2-3/h1-2H,10H2
InChI key
NPQBZKNXJZARBJ-UHFFFAOYSA-N
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service