Skip to Content
Merck
All Photos(1)

Key Documents

637513

Sigma-Aldrich

(S)-(+)-3-Fluoropyrrolidine hydrochloride

97%

Synonym(s):

(3S)-(+)-3-Fluoropyrrolidine hydrochloride, (3S)-3-Fluoropyrrolidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8FN · HCl
CAS Number:
Molecular Weight:
125.57
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D +8°, c = 1% in methanol

mp

183-187 °C (lit.)

SMILES string

Cl.F[C@H]1CCNC1

InChI

1S/C4H8FN.ClH/c5-4-1-2-6-3-4;/h4,6H,1-3H2;1H/t4-;/m0./s1

InChI key

LENYOXXELREKGZ-WCCKRBBISA-N

Application

(S)-(+)-3-Fluoropyrrolidine hydrochloride can be employed as a building block for the preparation of:
  • Fluorinated pyrrolidine derivatives of cyclohexylglycine amides as potential inhibitors for dipeptidyl peptidase IV.
  • Fluoropyrrolidine contaning N--ethylurea pyrazole derivatives as potential and selective inhibitors for Trypanosoma brucei and Trypanosoma cruzi parasitic causative agents.
  • Urea derivatives of (S)-2-(3,4-difluorophenyl)-5-(3-fluoro-N-pyrrolidylamido)benzothiazole as potent antitrypanosomal agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluoropyrrolidine amides as dipeptidyl peptidase IV inhibitors
Caldwell CG, et al.
Bioorganic & Medicinal Chemistry Letters, 14(5), 1265-1268 (2004)
Discovery of Potent N-Ethylurea Pyrazole Derivatives as Dual Inhibitors of Trypanosoma brucei and Trypanosoma cruzi
Varghese S, et al.
ACS Medicinal Chemistry Letters (2019)
Urea derivatives of 2-aryl-benzothiazol-5-amines: a new class of potential drugs for human African trypanosomiasis
Patrick DA, et al.
Journal of Medicinal Chemistry, 60(3), 957-971 (2017)
Charles G Caldwell et al.
Bioorganic & medicinal chemistry letters, 14(5), 1265-1268 (2004-02-26)
Amides derived from fluorinated pyrrolidines and 4-substituted cyclohexylglycine analogues have been prepared and evaluated as inhibitors of dipeptidyl dipeptidase IV (DP-IV). Analogues which incorporated (S)-3-fluoropyrrolidine showed good selectivity for DP-IV over quiescent cell proline dipeptidase (QPP). Compound 48 had good

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service