Skip to Content
Merck
All Photos(3)

Key Documents

637343

Sigma-Aldrich

Erbium(III) oxide

nanopowder, <100 nm particle size (BET), ≥99.9% trace metals basis

Synonym(s):

Dierbium trioxide, Erbia

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Er2O3
CAS Number:
Molecular Weight:
382.52
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: erbium

particle size

<100 nm (BET)

density

8.64 g/mL at 25 °C (lit.)

SMILES string

O=[Er]O[Er]=O

InChI

1S/2Er.3O

InChI key

VQCBHWLJZDBHOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Borm et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 36(5), 888-892 (1995-05-01)
The feasibility of postproduction neutron activation of an enteric-coated pancreatic enzyme preparation for in vivo gastric emptying studies has been investigated. During production of this multicomponent preparation, small amounts of 170Er-enriched erbium oxide, suitable for neutron activation, were added. Postproduction
Liu Peng et al.
Biological trace element research, 104(3), 275-284 (2005-06-03)
A microcalorimetric technique was used to evaluate the influence of Er3+ on Halobacterium halobium R1 growth. By means of a LKB-2277 Bioactivity Monitor ampoule method, we obtained the thermogenic curves of H. halobium R1 growth at 37 degrees C. In
Darya Radziuk et al.
Langmuir : the ACS journal of surfaces and colloids, 27(23), 14472-14480 (2011-10-26)
Ultrasound (20 kHz, 29 W·cm(-2)) is employed to form three types of erbium oxide nanoparticles in the presence of multiwalled carbon nanotubes as a template material in water. The nanoparticles are (i) erbium carboxioxide nanoparticles deposited on the external walls
Shixun Dai et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(1-3), 431-437 (2005-11-01)
(100-x)TeO(2)-xNb(2)O(5) (x=5-20) niobic tellurite glasses doped with 0.5 mol.% Er(2)O(3) were synthesized, and their thermal, mechanical, and spectroscopic properties were measured and compared to the properties of the typical 75TeO(2)-20ZnO-5Na(2)O (TZN) tellurite glass. The refractive index (n(d)), density (rho), and
Masao Kamimura et al.
Langmuir : the ACS journal of surfaces and colloids, 24(16), 8864-8870 (2008-07-26)
Infrared-to-visible upconversion phosphors (i.e., rare earth ion-doped Y2O3 nanoparticles (UNPs)) were synthesized by the homogeneous precipitation method. Because the charge on the erbium (Er) ion-doped Y2O3 (Y2O3:Er) NP (UNP1) surface is positive under neutral conditions, the UNP1 surface was electrostatically

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service