Skip to Content
Merck
All Photos(1)

Key Documents

611387

Supelco

Urea-13C,15N2 solution

NMR reference standard, 99 atom % 13C, 98 atom % 15N, 0.1 M in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 7 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CH415N2O
Molecular Weight:
63.03
UNSPSC Code:
12142201
NACRES:
NA.24

grade

NMR reference standard
analytical standard

isotopic purity

99 atom % 13C
98 atom % 15N

concentration

0.1 M in DMSO-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 7 in.

format

single component solution

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Indirect detection

Quantity

5 mm O.D. tube contains 0.700 mL.
Each pack contains 10 tubes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service