Skip to Content
Merck
All Photos(1)

Key Documents

607533

Sigma-Aldrich

2-Ketobutyric acid-4-13C,3,3,4,4,4-d5 sodium salt hydrate

48-70 atom % D (13CD3), 97 atom % D (CD2), 99 atom % 13C, 98% (CP)

Synonym(s):

α-Keto-butyric acid-4-13C,3,3,4,4,4-d5 sodium, α-Ketobutyric acid-4-13C,3,3,4,4,4-d5 sodium salt, 2-Oxobutanoic acid-4-13C.3,3,4,4,5-d5 sodium salt, Sodium α-ketobutyrate-4-13C,3,3,4,4,4-d5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CD3CD2COCO2Na · xH2O
Molecular Weight:
130.09 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C
48-70 atom % D (13CD3)
97 atom % D (CD2)

Quality Level

Assay

98% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

210 °C (dec.) (lit.)

mass shift

M+6

SMILES string

O.[Na+].[2H][13C]([2H])([2H])C([2H])([2H])C(=O)C([O-])=O

InChI

1S/C4H6O3.Na.H2O/c1-2-3(5)4(6)7;;/h2H2,1H3,(H,6,7);;1H2/q;+1;/p-1/i1+1D3,2D2;;

InChI key

PVLJVKQSCAHPPR-RWBNLZSCSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.

We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.

We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.

We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service