Skip to Content
Merck
All Photos(1)

Key Documents

588393

Sigma-Aldrich

Cetyl(pyridinium-d5) chloride monohydrate

98 atom % D, 98% (CP)

Synonym(s):

Hexadecyl-(pyridinium-d5)-chloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D5H33ClN·H2O
Molecular Weight:
363.03
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

mp

81 °C (lit.)

mass shift

M+5

SMILES string

O.[Cl-].[2H]c1c([2H])c([2H])[n+](CCCCCCCCCCCCCCCC)c([2H])c1[2H]

InChI

1S/C21H38N.ClH.H2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-22-20-17-15-18-21-22;;/h15,17-18,20-21H,2-14,16,19H2,1H3;1H;1H2/q+1;;/p-1/i15D,17D,18D,20D,21D;;

InChI key

NFCRBQADEGXVDL-UGIAOSMXSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qihao Zhang et al.
American journal of translational research, 12(5), 2062-2070 (2020-06-09)
The quantity and function of osteoblasts require continuous osteogenic differentiation of bone marrow mesenchymal stem cells. Recent evidence suggests that microRNAs (miRNAs) act as important post-transcriptional regulators in a wide range of biological processes, including osteoblastic differentiation. Quercetin has also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service