Skip to Content
Merck
All Photos(1)

Key Documents

558508

Sigma-Aldrich

2′-Hydroxy-4,4′,6′-trimethoxychalcone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH=CHCOC6H2(OCH3)2OH
CAS Number:
Molecular Weight:
314.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%

Quality Level

Assay

98%

form

solid

mp

112-116 °C (lit.)

functional group

ketone

SMILES string

COc1ccc(\C=C\C(=O)c2c(O)cc(OC)cc2OC)cc1

InChI

1S/C18H18O5/c1-21-13-7-4-12(5-8-13)6-9-15(19)18-16(20)10-14(22-2)11-17(18)23-3/h4-11,20H,1-3H3/b9-6+

InChI key

CGIBCVBDFUTMPT-RMKNXTFCSA-N

Application

2′-Hydroxy-4,4′,6′-trimethoxychalcone may be used to synthesize 2′,2”′-dihydroxy-4,4′,4′′,4”′,6′,6′′′-hexamethoxy[5′,5′′′]bichalcone and 3′-bromo-4,4′,6′-trimethoxy-2′-hydroxychalcone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical conversion of artemisinic acid in desoxyartemisinin.
Jung M, et al.
The Journal of Organic Chemistry, 51(26), 5417-5419 (1986)
Regioselective bromination of organic substrates by tetrabutylammonium bromide promoted by V2O5-H2O2: An environmentally favorable synthetic protocol.
Bora U, et al.
Organic Letters, 2(3), 247-249 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service