Skip to Content
Merck
All Photos(2)

Key Documents

556378

Sigma-Aldrich

Polyaniline (emeraldine base)

average Mw ~20,000

Synonym(s):

Emeraldine base polyaniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw ~20,000

solubility

m-cresol: soluble
DMAC: soluble
DMF: soluble
DMSO: soluble
NMP: soluble

λmax

328 nm

SMILES string

Brc1nn[nH]c1C.Cl

InChI

1S/C3H4BrN3.ClH/c1-2-3(4)6-7-5-2;/h1H3,(H,5,6,7);1H

InChI key

LYCWKZFKTMHXQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyaniline (PAni) emeraldine salt can be prepared by protonation of PAni emeraldine base (PAni-EB). PAni-EB forms at pH>7. Its mechanism against corrosive agents has been explained in a study.
Polyaniline (emeraldine base) (PANI-EB) is a conductive polymer that can be chemically synthesized from aniline by oxidative polymerization of ammonium peroxydisulfate in acidic solution. It is widely used in electronics and optoelectronic applications.

Application

PANI-EB is a conjugating polymer that finds applications in organic light emitting diodes (OLEDs), anticorrosive coatings, chemical sensors and solar cells.

Features and Benefits

Undoped form of conducting polymer (non-conductive).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anticorrosion performances of epoxy coatings modified with polyaniline: A comparison between the emeraldine base and salt forms
Armelin E, et al.
Progress in Organic Coatings, 65(1), 88-93 (2009)
Polyaniline nanofiber gas sensors: examination of response mechanisms.
Virji S, et al.
Nano Letters, 4(3), 491-496 (2004)
Polyaniline/TiO2 solar cells.
Liu Z, et al.
Synthetic Metals, 156(9-10), 721-723 (2006)
Emeraldine Base Polyaniline as an Alternative to Poly(3,4-ethylenedioxythiophene) as a Hole-Transporting Layer
Higgins RWT, et al.
Advances in Functional Materials, 11(6), 407-402 (2001)
Polyaniline nanofibers: facile synthesis and chemical sensors
Huang J, et al.
Journal of the American Chemical Society, 125(2), 314-315 (2003)

Articles

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service