Skip to Content
Merck
All Photos(1)

Key Documents

540293

Sigma-Aldrich

Tetraalkylammonium carbonate, polymer-bound

macroporous, 40-90 mesh, extent of labeling: 2.5-3.5 mmol/g loading

Synonym(s):

MP-carbonate resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

form

macroporous

Quality Level

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

2.5-3.5 mmol/g loading

particle size

40-90 mesh

SMILES string

[O-]C([O-])=O.C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.CC[N+](CC)(CC)Cc3ccccc3.CC[N+](CC)(CC)Cc4ccccc4

InChI

1S/2C13H22N.C10H10.C8H8.CH2O3/c2*1-4-14(5-2,6-3)12-13-10-8-7-9-11-13;1-3-9-5-7-10(4-2)8-6-9;1-2-8-6-4-3-5-7-8;2-1(3)4/h2*7-11H,4-6,12H2,1-3H3;3-8H,1-2H2;2-7H,1H2;(H2,2,3,4)/q2*+1;;;/p-2

InChI key

RBIYTBCFDIQNIC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Polymer-bound tetraalkylammonium carbonate is a polymer-supported base used for scavenging acidic molecules. It can be used in polymer-assisted solution-phase (PASP) Suzuki reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer-assisted solution-phase (PASP) Suzuki couplings employing an anthracene-tagged palladium catalyst.
Lan P, et al.
The Journal of Organic Chemistry, 68(25), 9678-9686 (2003)
S R Stauffer et al.
Journal of combinatorial chemistry, 2(4), 318-329 (2000-07-13)
Most ligands for the estrogen receptor (ER) are not well suited for synthesis by combinatorial means, because their construction involves a series of carbon-carbon bond forming reactions that are not uniformly high yielding. In previous work directed to overcoming this
Parlow, J. et al.
Tetrahedron Letters, 38, 7959-7959 (1999)
Recent applications of polymer-supported reagents and scavengers in combinatorial, parallel, or multistep synthesis.
Thompson L A.
Current Opinion in Chemical Biology, 4(3), 324-337 (2000)
Ley, S.V. et al.
J. Chem. Soc., Perkin Trans., 1251-1251 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service