Skip to Content
Merck
All Photos(1)

Key Documents

540072

Sigma-Aldrich

(R)-(−)-Epichlorohydrin

99%

Synonym(s):

(R)-(−)-2-(Chloromethyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
Beilstein:
1420785
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.29 (vs air)

vapor pressure

10 mmHg ( 16.6 °C)

Assay

99%

optical activity

[α]20/D −34°, c = 1 in methanol

optical purity

ee: 98% (GLC)

autoignition temp.

772 °F

expl. lim.

21 %

refractive index

n20/D 1.438 (lit.)

bp

114 °C (lit.)

density

1.180 g/mL at 20 °C (lit.)

SMILES string

ClC[C@H]1CO1

InChI

1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2/t3-/m0/s1

InChI key

BRLQWZUYTZBJKN-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Building block for the synthesis of a key intermediate in the synthesis of stable PGI2 analogue UT-15.
Used in the synthesis of chiral morpholines and dioxanes via a Mitsunobu diol-cyclization. Chiral building block in the enantioselective synthesis of trans-2,4-disubstituted piperidines.

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Robert M Moriarty et al.
The Journal of organic chemistry, 69(6), 1890-1902 (2004-04-03)
A general and novel solution to the synthesis of biologically important stable analogues of prostacyclin PGI(2), namely benzindene prostacyclins, has been achieved via the stereoselective intramolecular Pauson-Khand cyclization (PKC). This work illustrates for the first time the synthetic utility and
Goss S Kauffman et al.
The Journal of organic chemistry, 71(23), 8975-8977 (2006-11-04)
A strategy for the enantioselective synthesis of trans-2,4-disubstituted piperidines is proposed and applied to the preparation of IS811, a potent CCR3 antagonist. The C2 stereocenter is derived from commercial (R)-epichlorohydrin, while the C4 stereocenter is installed via diastereoselective hydrogenation of
Synlett, 2151-2151 (2006)
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 16(10), 5720-5732 (2008-04-15)
The risk of the presence of haloacetic acids in drinking water as chlorination by-products and the shortage of experimental mutagenicity data for most of them requires a research work. This paper describes a QSAR model to predict direct mutagenicity for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service